Skip to Content
Merck
All Photos(1)

Key Documents

383813

Sigma-Aldrich

Zinc 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine

Synonym(s):

1,4,8,11,15,18,22,25-Octabutoxyphthalocyanine zinc salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$156.00

$156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C64H80N8O8Zn
CAS Number:
Molecular Weight:
1154.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$156.00


Check Cart for Availability

Request a Bulk Order

mp

185 °C (dec.) (lit.)

λmax

736 nm

SMILES string

CCCCOc1ccc(OCCCC)c2c3nc(nc4n5[Zn]n6c(n3)c7c(OCCCC)ccc(OCCCC)c7c6nc8nc(nc5c9c(OCCCC)ccc(OCCCC)c49)c%10c(OCCCC)ccc(OCCCC)c8%10)c12

InChI

1S/C64H80N8O8.Zn/c1-9-17-33-73-41-25-26-42(74-34-18-10-2)50-49(41)57-65-58(50)70-60-53-45(77-37-21-13-5)29-30-46(78-38-22-14-6)54(53)62(67-60)72-64-56-48(80-40-24-16-8)32-31-47(79-39-23-15-7)55(56)63(68-64)71-61-52-44(76-36-20-12-4)28-27-43(75-35-19-11-3)51(52)59(66-61)69-57;/h25-32H,9-24,33-40H2,1-8H3;/q-2;+2

InChI key

DKGNTSAYUCTYJI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Fabris et al.
Journal of photochemistry and photobiology. B, Biology, 39(3), 279-284 (1997-07-01)
Zn(II)-octapentyl-phthalocyanine (ZnOPPc), incorporated into a Cremophor oil emulsion, was found to be a highly selective tumour-targeting agent (50-fold larger concentration in the tumour than in the peritumoral tissue) when injected at a dose of 1.46 mumol (kg body weight)-1 in
W E Ford et al.
Photochemistry and photobiology, 50(3), 277-282 (1989-09-01)
Triplet-state properties of 1,4,8,11,15,18,22,25-octa-n-butoxyphthalocyanine and its zinc derivative were determined for the first time. The T1 state of the metal-free phthalocyanine was characterized by a short lifetime (tau T = 17 microseconds) and low quantum yield (phi T = 0.095)
G Jori et al.
Journal of photochemistry and photobiology. B, Biology, 43(3), 181-185 (1998-08-27)
Zn(II)-phthalocyanines (ZnPc) and its octapentyl (ZnOPPc) and octadecyl (ZnODPc) derivatives have been intravenously injected at a dose of 1.46 mumol/kg into female Balb/c mice bearing an intramuscularly transplanted MS-2 fibrosarcoma. Pharmacokinetic studies show that in all cases the maximal concentration
C Ometto et al.
British journal of cancer, 74(12), 1891-1899 (1996-12-01)
1,4,8,11,15,18,22,25-Octadecylphthalocyaninato zinc(II), ZnODPc, incorporated into a Cremophor emulsion, was assayed for its pharmacokinetic and phototherapeutic properties in Balb/c mice bearing an intramuscularly transplanted MS-2 fibrosarcoma. The phthalocyanine was injected intravenously (i.v.) in three doses, i.e. 1.46, 0.73 and 0.37 mumol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service