Skip to Content
Merck
All Photos(1)

Key Documents

382639

Sigma-Aldrich

4-(Hydroxymethyl)benzoic acid

99%

Synonym(s):

p-(Hydroxymethyl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$38.50
10 G
$206.00

$38.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$38.50
10 G
$206.00

About This Item

Linear Formula:
HOCH2C6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2690023
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$38.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

reaction suitability

reagent type: cross-linking reagent

mp

182-185 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

O=C(O)C1=CC=C(CO)C=C1

InChI

1S/C8H8O3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

WWYFPDXEIFBNKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsunehisa Hirose et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-05)
The retention behavior of a wide variety of stationary phases was compared in supercritical fluid chromatography (SFC) and normal-phase high-performance liquid chromatography (NP-HPLC). We also attempted to elucidate the retention behavior in SFC by investigating the selectivity of the different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service