Skip to Content
Merck
All Photos(1)

Key Documents

382485

Sigma-Aldrich

5-Methoxy-2-benzimidazolethiol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$58.90

$58.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$58.90

About This Item

Empirical Formula (Hill Notation):
C8H8N2OS
CAS Number:
Molecular Weight:
180.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

mp

261-263 °C (lit.)

SMILES string

COc1ccc2[nH]c(S)nc2c1

InChI

1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4H,1H3,(H2,9,10,12)

InChI key

KOFBRZWVWJCLGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methoxy-2-benzimidazolethiol is a hetercocyclic building block. Iodimetric quantification of 5-methoxy-2-benzimidazolethiol in basic medium has been investigated.[1]

Application

5-Methoxy-2-benzimidazolethiol may be used in the preparation of compounds present as contaminants in omeprazole, such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole.[2] It may be used in the synthesis of tris[μ-1,2-bis(diphenylphosphino)ethane]-1:2κ2P:P′;1:3κ2P:P′;2:3κ2P:P′-di-μ-bromido-1:2κ4Br:Br-bromido-3κBr-tricopper(I) acetone hemisolvate.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

286.9 °F - closed cup

Flash Point(C)

141.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Juan Shi
Acta crystallographica. Section E, Structure reports online, 64(Pt 11), m1411-m1411 (2008-01-01)
In the crystal structure of the title compound, [Cu(3)Br(3)(C(26)H(24)P(2))(3)]·0.5CH(3)COCH(3), two of the Cu centers are bridged by two bromide anions forming a Cu(μ-Br)(2)Cu core, which is further bridged by a 1,2-bis-(diphenyl-phosphino)-ethane (dppe) ligand. The third Cu center is terminally bound
Preparation of Two Impurities of Omeprazole.
Hao L-H, et al.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 2, 008-008 (2009)
Iodimetric determination of 2-benzimidazolethiols and 2-benzothiazoletiols.
Ciesielski W and Krenc A.
Chem. Anal. (Warsaw), 49(4), 577-577 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service