Skip to Content
Merck
All Photos(1)

Documents

381578

Sigma-Aldrich

1-Butyl-3-sulfanilylurea

98%

Synonym(s):

Carbutamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO2NHCONH(CH2)3CH3
CAS Number:
Molecular Weight:
271.34
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

mp

143-145 °C (lit.)

SMILES string

CCCCNC(=O)NS(=O)(=O)c1ccc(N)cc1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service