Skip to Content
Merck
All Photos(3)

Key Documents

381454

Sigma-Aldrich

Butyl 3-mercaptopropionate

98%

Synonym(s):

Butyl 3-mercaptopropanoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$79.00
500 ML
$199.00

$79.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$79.00
500 ML
$199.00

About This Item

Linear Formula:
HSCH2CH2CO2(CH2)3CH3
CAS Number:
Molecular Weight:
162.25
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$79.00


Available to ship on15 April 2025Details


Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.457 (lit.)

bp

101 °C/12 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)CCS

InChI

1S/C7H14O2S/c1-2-3-5-9-7(8)4-6-10/h10H,2-6H2,1H3

InChI key

MGFFVSDRCRVHLC-UHFFFAOYSA-N

General description

Butyl 3-mercaptopropionate (3MPA) is a monofunctional thiol that can be used as a cross-linker and chain transferring agent for controlling the molecular weight of the polymer. It can be used in the thiolene photopolymerization.[1][2][3]

Application

3MPA can be used as a ligand which can functionalize the quantum dots for the development of high luminescence light emitting diodes.[4] It can also be used as a crosslinking monomeric unit for the preparation of thiol-acrylate based photopolymers.[5]
Used as a reactant for thiol-yne photopolymerizations to form highly-cross-linked networks.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Initiation and kinetics of thiol-ene photopolymerizations without photoinitiators
Cramer NB, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 42(22), 5817-5826 (2004)
High luminescence efficiency white light emitting diodes based on surface functionalized quantum dots dispersed in polymer matrices
Yoon C, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 428(22), 86-91 (2013)
Development of a poly (methyl methacrylate-co-n-butyl methacrylate) copolymer binder system
Vail NK, et al.
Journal of Applied Polymer Science, 52(6), 789-812 (1994)
Effects of neighboring sulfides and pH on ester hydrolysis in thiol-acrylate photopolymers
Rydholm AE, et al.
Acta Biomaterialia, 3(4), 449-455 (2007)
Thiol- Yne photopolymerizations: novel mechanism, kinetics, and step-growth formation of highly cross-linked networks
Fairbanks BD, et al.
Macromolecules, 42(1), 211-217 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service