Skip to Content
Merck
All Photos(2)

Key Documents

381292

Sigma-Aldrich

Antimony(III) fluoride

powder, 99.8% trace metals basis

Synonym(s):

Antimony trifluoride, Trifluorostibine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$233.00
500 G
$902.00

$233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$233.00
500 G
$902.00

About This Item

Linear Formula:
SbF3
CAS Number:
Molecular Weight:
178.76
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$233.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.8% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: antimony

impurities

≤2500.0 ppm Trace Metal Analysis

mp

235 °C (dec.) (lit.)

density

4.38 g/mL at 25 °C (lit.)

SMILES string

F[Sb](F)F

InChI

1S/3FH.Sb/h3*1H;/q;;;+3/p-3

InChI key

GUNJVIDCYZYFGV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Structure and Properties of the Molecular Complex of Antimony(III) Fluoride with γ-Glycine: The paper details the structural type and properties of the antimony(III) fluoride complex with γ-glycine, contributing to the understanding of complexation behavior in these systems (Voit et al., 2019).

  • Crystal Structure and Ion Mobility in the Antimony(III) Fluoride Complex with Glycine: The study investigates the synthesis and ion mobility in antimony(III) fluoride complex with glycine, contributing to the field of coordination chemistry (Makarenko et al., 2018).

Other Notes

Contains lead

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service