Skip to Content
Merck
All Photos(2)

Key Documents

379794

Sigma-Aldrich

Sodium tetrachloroaurate(III) hydrate

99.998% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$454.00

$454.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$454.00

About This Item

Linear Formula:
NaAuCl4 · xH2O
CAS Number:
Molecular Weight:
361.77 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

$454.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.998% trace metals basis

form

solid

impurities

≤30.0 ppm Trace Metal Analysis

SMILES string

O.[Na+].Cl[Au-](Cl)(Cl)Cl

InChI

1S/Au.4ClH.Na.H2O/h;4*1H;;1H2/q+3;;;;;+1;/p-4

InChI key

VTMCVHXKFJBNKC-UHFFFAOYSA-J

Related Categories

Application

A water/alcohol soluble Au(I) complex formed from NaAuCl4 has potential for application in chemotherapeutics.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas E. Berning et al.
Inorganic chemistry, 36(13), 2765-2769 (1997-06-18)
The reactions of water-soluble bisphosphines (HOH(2)C)(2)PC(6)H(4)P(CH(2)OH)(2) (1) and (HOH(2)C)(2)PCH(2)CH(2)P(CH(2)OH)(2) (2) with NaAuCl(4), in aqueous media, or AuClPPh(3), in biphasic media (aqueous/organic), produced the water/alcoholic-soluble Au(I) complexes [Au{(HOH(2)C)(2)PC(6)H(4)P(CH(2)OH)(2)}(2)]Cl (3), [Au{(HOH(2)C)(2)PCH(2)CH(2)P(CH(2)OH)(2)}(2)]Cl (4), and [Au(2){(HOH(2)C)(2)PCH(2)CH(2)P(CH(2)OH)(2)}(2)]Cl(2) (5) in near quantitative yields. Stability and cysteine-challenge

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service