Skip to Content
Merck
All Photos(1)

Key Documents

379204

Sigma-Aldrich

Brilliant Blue R

Dye content 50 %

Synonym(s):

Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 50%

λmax

588 nm

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For detection of protein bands following electrophoresis.

Legal Information

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montse Olivé et al.
Neuromuscular disorders : NMD, 18(12), 929-933 (2008-10-25)
A 27-year-old woman of Moldavian origin presented at the age of 15 with progressive proximal limb weakness and painful cramps in her calf muscles. Clinical examination revealed prominent muscle weakness in proximal muscles of the lower extremities and distal anterior
Reiner Westermeier
Proteomics, 6 Suppl 2, 61-64 (2006-10-13)
In spite of the high sensitivity of silver staining and the wide dynamic range of various fluorescent detection methods, Coomassie Brilliant Blue staining is still the most widely used protein detection technique for proteins separated by polyacrylamide gel electrophoresis. There
Zhang Xin-Guo Chen Jian-Hua et al.
Journal of chromatographic science, 50(9), 820-825 (2012-06-22)
A human serum albumin and Thymosin α1 (HSA-Tα1) fusion protein was designed and over-expressed in Pichia pastoris. To purify the fusion protein, a new native preparative electrophoresis system that involved a modified device with a sample receiving chamber, and an
K Kubo
Analytical biochemistry, 213(2), 200-205 (1993-09-01)
In sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) according to the method of Weber and Osborn, protein bands are often distorted by tailing at both ends, and the phenomenon is often called "edge tailing." This was eliminated by adding glycerol at
Yong-Hak Kim et al.
Archives of biochemistry and biophysics, 494(2), 159-165 (2009-12-01)
We studied the decolorization of malachite green (MG) by the fungus Cunninghamella elegans. The mitochondrial activity for MG reduction was increased with a simultaneous increase of a 9-kDa protein, called CeCyt. The presence of cytochrome c in CeCyt protein was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service