Skip to Content
Merck
All Photos(2)

Key Documents

377619

Sigma-Aldrich

D-Threitol

99%

Synonym(s):

(2R,3R)-1,2,3,4-Butanetetrol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$167.00

$167.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$167.00

About This Item

Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
Beilstein:
1719752
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

$167.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

optical activity

[α]20/D −14°, c = 2 in ethanol

mp

88-90 °C (lit.)

SMILES string

OC[C@@H](O)[C@H](O)CO

InChI

1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

UNXHWFMMPAWVPI-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Bravo et al.
Carbohydrate research, 336(2), 83-97 (2001-11-02)
Differently protected erythro and threo furanoid glycals were synthesized by selenoxide elimination when phenyl 1-selenoglycosides were treated in oxidizing conditions (tBuOOH, Ti(O(i)Pr)(4), Et(2)(i)PrN). The phenyl 1-selenoglycosides were obtained from methyl 2-deoxy-D-erythro-pentofuranoside by protection of the primary hydroxyl or both hydroxyls
Justyna Wojno et al.
ACS chemical biology, 7(5), 847-855 (2012-02-14)
Invariant natural killer T (iNKT) cells are restricted by the non-polymorphic MHC class I-like protein, CD1d, and activated following presentation of lipid antigens bound to CD1d molecules. The prototypical iNKT cell agonist is α-galactosyl ceramide (α-GalCer). CD1d-mediated activation of iNKT
N Tbeur et al.
Carbohydrate research, 329(2), 409-422 (2000-12-16)
A supported liquid membrane (SLM) containing a resorcinarene carrier has been used for the selective transport of erythritol, threitol, ribitol and xylitol from concentrated (1.0-0.01 M) aqueous solutions. The membrane is made of a microporous polytetrafluoroethylene film impregnated with a
J M Borggreven et al.
Caries research, 26(2), 84-88 (1992-01-01)
Some newly designed surface-active phosphates were investigated with respect to their capacity to inhibit caries lesion formation in vitro. On the labial or buccal surfaces of sound human third molars, windows were prepared for investigation. One half of each window
Leonardo Silva Santos et al.
Rapid communications in mass spectrometry : RCM, 20(14), 2104-2108 (2006-06-13)
Recently, it has been proposed (M. Claeys et al., Science 2004; 303: 1173) that the atmospheric OH-radical-mediated photooxidation of isoprene is a source of two major secondary organic aerosol (SOA) components, that is, 2-methylthreitol and 2-methylerythritol. These diastereoisomeric tetrols, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service