Skip to Content
Merck
All Photos(2)

Key Documents

377538

Sigma-Aldrich

Heptyltriphenylphosphonium bromide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$186.00

$186.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
50 G
$186.00

About This Item

Linear Formula:
CH3(CH2)6P(C6H5)3Br
CAS Number:
Molecular Weight:
441.38
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$186.00


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

173-176 °C (lit.)

functional group

phosphine

SMILES string

[Br-].CCCCCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C25H30P.BrH/c1-2-3-4-5-15-22-26(23-16-9-6-10-17-23,24-18-11-7-12-19-24)25-20-13-8-14-21-25;/h6-14,16-21H,2-5,15,22H2,1H3;1H/q+1;/p-1

InChI key

WCZSOHSGMBVYFW-UHFFFAOYSA-M

Application

Reactant for:
  • Stereospecific rearrangement of epoxides to quaternary carbaldehydes
  • Synthesis of the male pheromone of two species of termite Zootermopsis
  • Regio- and stereoselective TEMPO oxidation
  • Diastereoselective synthesis of climacostol for antitumor activity
  • Enantioselective synthesis of chiral vinylphosphonate and phosphonate analogs of immunosuppressive agent FTY720
  • Preparation of embelin derivatives with XIAP inhibitory and anticancer activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tejia Zhang et al.
Journal of lipid research, 56(8), 1501-1510 (2015-06-11)
Apoptosis is an intricately regulated cellular process that proceeds through different cell type- and signal-dependent pathways. In the mitochondrial apoptotic program, mitochondrial outer membrane permeabilization by BCL-2 proteins leads to the release of apoptogenic factors, caspase activation, and cell death.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service