Skip to Content
Merck
All Photos(2)

Key Documents

375942

Sigma-Aldrich

Diisopropylsilyl bis(trifluoromethanesulfonate)

96%

Synonym(s):

Trifluoromethanesulfonic acid diisopropylsilylene ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$204.00

$204.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$204.00

About This Item

Linear Formula:
(CF3SO3)2Si[CH(CH3)2]2
CAS Number:
Molecular Weight:
412.40
Beilstein:
3567500
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$204.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.382 (lit.)

bp

85-86 °C/2 mmHg (lit.)

density

1.396 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

CC(C)[Si](OS(=O)(=O)C(F)(F)F)(OS(=O)(=O)C(F)(F)F)C(C)C

InChI

1S/C8H14F6O6S2Si/c1-5(2)23(6(3)4,19-21(15,16)7(9,10)11)20-22(17,18)8(12,13)14/h5-6H,1-4H3

InChI key

BHZPNBNSRVSTBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service