Skip to Content
Merck
All Photos(1)

Key Documents

375764

Sigma-Aldrich

C-Methylcalix[4]resorcinarene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$81.40

$81.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$81.40

About This Item

Empirical Formula (Hill Notation):
C32H32O8
CAS Number:
Molecular Weight:
544.59
Beilstein:
9110521
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$81.40


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

>300 °C (lit.)

SMILES string

[H]C1(C)c2cc(c(O)cc2O)C([H])(C)c3cc(c(O)cc3O)C([H])(C)c4cc(c(O)cc4O)C([H])(C)c5cc1c(O)cc5O

InChI

1S/C32H32O8/c1-13-17-5-19(27(35)9-25(17)33)14(2)21-7-23(31(39)11-29(21)37)16(4)24-8-22(30(38)12-32(24)40)15(3)20-6-18(13)26(34)10-28(20)36/h5-16,33-40H,1-4H3

InChI key

WGDNYTQTRISCMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of selective receptors for aqueous mercury(II) and complexation with copper(II) and silver(I)
  • Synthesis of nonchemically amplified molecular resists used as a dissolution promotor
  • Reactions with glycidyl Ph ether used for thermal curing of epoxy resin
  • Mannich reactions
  • Mediation of prenucleation and coalescence of cobalt nanoclusters
  • Atom transfer radical polymerization for synthesis of a star polymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed K Abd El-Rahman et al.
Talanta, 221, 121409-121409 (2020-10-21)
Choline (Ch+), is a vitamin-like essential water-soluble organic micronutrient. The US-FDA requires that infant formula not made from cow's milk must be supplemented with Ch+. Direct determination of Ch+ in milk powders and infant formulas is a challenging task due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service