Skip to Content
Merck
All Photos(2)

Key Documents

374989

Sigma-Aldrich

Triisobutylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]3N
CAS Number:
Molecular Weight:
185.35
Beilstein:
1698950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.423 (lit.)

bp

192-193 °C/770 mmHg (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CC(C)CN(CC(C)C)CC(C)C

InChI

1S/C12H27N/c1-10(2)7-13(8-11(3)4)9-12(5)6/h10-12H,7-9H2,1-6H3

InChI key

IIFFFBSAXDNJHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triisobutylamine is a sterically hindered amine. The rate constants for H-atom abstraction from triisobutylamine has been evaluated.[1]
Triisobutylamine is an organic base.[2] Solubility and liquid-liquid equilibria of binary mixtures of triisobutylamine and water has been evaluated.[3]

Application

Triisobutylamine is suitable to develop second generation ionic liquid matrices for MALDI-MS of peptides, proteins and carbohydrates.[4] It may be used in the separation of cis-fatty acid salts and and trans-fatty acid (elaidic acid) salts using a new organic solvent nanofiltration membrane based on polydicyclopentadiene.[5] It may be used as base in the synthesis of acroyl ammonium derivatives.[6]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

IUPAC-NIST Solubility Data Series. 96. Amines with Water Part 2. C7-C24 Aliphatic Amines.
Goral M, et al.
Journal of Physical and Chemical Reference Data, 41(4), 043107-043107 (2012)
Synthesis of a Fused ?-Lactone via Pyridine-Assisted [2+ 2] Cycloaddition.
Reddy LR and Corey EJ.
Synfacts, 06, 0550-0550 (2006)
Nicholas A Isley et al.
Journal of the American Chemical Society, 135(47), 17707-17710 (2013-11-15)
New technology has been developed that enables Suzuki-Miyaura couplings involving widely utilized MIDA boronates to be run in water as the only medium, mainly at room temperature. The protocol is such that no organic solvent is involved at any stage;
Jeffrey A Crank et al.
Journal of the American Society for Mass Spectrometry, 20(10), 1790-1800 (2009-08-12)
Second generation ionic liquid matrices are developed, examined, and tested. They have shown a wide mass detection range (<1000 Da to >270,000 Da) for proteins and peptides with greater S/N ratios than solid matrices. These ionic liquid matrices also exhibit
Michela Salamone et al.
Organic letters, 13(2), 260-263 (2010-12-15)
The rate constants for H-atom abstraction (k(H)) from 1,4-cyclohexadiene (CHD), triethylamine (TEA), triisobutylamine (TIBA), and DABCO by the cumyloxyl (CumO(•)) and benzyloxyl (BnO(•)) radicals were measured. Comparable k(H) values for the two radicals were obtained in their reactions with CHD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service