374326
2,5-Diphenyl-1,6,6aλ4-trithiapentalene
97%
Synonym(s):
2,5-Diphenyl[1,2]dithiolo[1,5-b][1,2]dithiole-7-SIV
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H12S3
CAS Number:
Molecular Weight:
312.47
Beilstein:
19419
MDL number:
Assay
97%
SMILES string
c1ccc(cc1)-c2cc3cc(ss3s2)-c4ccccc4
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service