Skip to Content
Merck
All Photos(1)

Key Documents

37347

Sigma-Aldrich

O-(2-Oxo-1(2H)pyridyl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate

≥99.0% (HPLC)

Synonym(s):

O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N′-N′-tetramethyluronium tetrafluoroborate, TPTU

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$255.00
25 G
$649.00

$255.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$255.00
25 G
$649.00

About This Item

Empirical Formula (Hill Notation):
C10H16BF4N3O2
CAS Number:
Molecular Weight:
297.06
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$255.00


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC)

reaction suitability

reaction type: Coupling Reactions

mp

140 °C (dec.) (lit.)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CN(C)C(\ON1C=CC=CC1=O)=[N+](/C)C

InChI

1S/C10H16N3O2.BF4/c1-11(2)10(12(3)4)15-13-8-6-5-7-9(13)14;2-1(3,4)5/h5-8H,1-4H3;/q+1;-1

InChI key

CZQGINAUZYECAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Amidation[1]
Peptide Coupling[2][3]
Esterification of nucleosides to solid phase supports for oligonucleoside synthesis[4]

Other Notes

Coupling reagent for peptide synthesis, especially suited for segment condensation with little racemization[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Huntley et al.
Nucleosides, nucleotides & nucleic acids, 20(4-7), 731-733 (2001-09-21)
The synthesis of 1-(beta-D-ribofuranosyl)pyridin-2-one-3-carboxylic acid and the 3-carboxamide as well as a short series of 3N-carboxamides, prepared by TPTU/HOBt coupling of primary amines with 1-(beta-D-ribofuranosyl)pyridin-2-one-3-carboxylic acid, and their evaluation as anti-infective agents is described.
R T Pon et al.
Bioconjugate chemistry, 10(6), 1051-1057 (1999-11-24)
Nucleosides can be esterified to solid-phase supports using uronium or phosphonium coupling reagents and a coupling additive, such as 1-hydroxybenzotriazole (HOBT), 7-aza-1-hydroxybenzotriazole (HOAT), N-methylimidazole (NMI), or 4-(dimethylamino)pyridine (DMAP). However, DMAP was far superior to other additives and high nucleoside loadings
Tomohisa Sawada et al.
Journal of the American Chemical Society, 133(19), 7336-7339 (2011-04-28)
Artificial mimicry of α-helices offers a basis for development of protein-protein interaction antagonists. Here we report a new type of unnatural peptidic backbone, containing α-, β-, and γ-amino acid residues in an αγααβα repeat pattern, for this purpose. This unnatural
Knorr, R, et al.
Tetrahedron Letters, 30, 1927-1927 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service