Skip to Content
Merck
All Photos(2)

Key Documents

372110

Sigma-Aldrich

4-sec-Butyl-2,6-di-tert-butylphenol

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$142.00

$142.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$142.00

About This Item

Linear Formula:
C2H5CH(CH3)C6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
262.43
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$142.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

bp

141-142 °C/10 mmHg (lit.)

mp

25 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

CCC(C)c1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C18H30O/c1-9-12(2)13-10-14(17(3,4)5)16(19)15(11-13)18(6,7)8/h10-12,19H,9H2,1-8H3

InChI key

BFZOTKYPSZSDEV-UHFFFAOYSA-N

General description

Synthesis of 4-sec-butyl-2,6-di-tert-butylphenol from 4-(sec-butyl) phenol and isobutene using sulfuric resin as catalyst, is reported.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 4-(Sec-butyl)-2, 6-di-tert-butylphenol.
ZANG Y-L, et al.
Fine Chemicals / ????, 1, 018-018 (2012)
Paula Vera et al.
Talanta, 188, 750-762 (2018-07-22)
Migration of non volatile compounds from twenty six PP films used as food contact materials has been studied in four simulants (ethanol 95% and 10%, acetic acid 3% and Tenax ®) and analyzed by UPLC-MS/QTOF. Seventy six compounds have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service