Skip to Content
Merck
All Photos(2)

Key Documents

367869

Sigma-Aldrich

Brucine hydrate

technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$370.00

$370.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$370.00

About This Item

Empirical Formula (Hill Notation):
C23H26N2O4 · xH2O
CAS Number:
Molecular Weight:
394.46 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$370.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

technical grade

form

solid

optical activity

[α]20/D −67°, c = 1.3 in ethanol

mp

177-180 °C (lit.)

functional group

ether

SMILES string

[H]O[H].[H][C@@]12CC(=O)N3c4cc(OC)c(OC)cc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H]

InChI

1S/C23H26N2O4.H2O/c1-27-16-8-14-15(9-17(16)28-2)25-20(26)10-18-21-13-7-19-23(14,22(21)25)4-5-24(19)11-12(13)3-6-29-18;/h3,8-9,13,18-19,21-22H,4-7,10-11H2,1-2H3;1H2/t13-,18-,19-,21-,22-,23+;/m0./s1

InChI key

IEJIBKWJYHZFNW-SUJBTXFYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service