Skip to Content
Merck
All Photos(1)

Key Documents

366331

Sigma-Aldrich

Trimethyl(phenyl)tin

98%

Synonym(s):

Phenyltrimethylstannane, Phenyltrimethyltin, Trimethylmonophenyltin, Trimethylphenylstannane, Trimethylstannylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$66.30
5 G
$225.00

$66.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$66.30
5 G
$225.00

About This Item

Linear Formula:
C6H5Sn(CH3)3
CAS Number:
Molecular Weight:
240.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$66.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

reaction suitability

core: tin

refractive index

n20/D 1.5357 (lit.)

bp

88 °C/16 mmHg (lit.)

mp

−51 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

SMILES string

C[Sn](C)(C)c1ccccc1

InChI

1S/C6H5.3CH3.Sn/c1-2-4-6-5-3-1;;;;/h1-5H;3*1H3;

InChI key

COHOGNZHAUOXPA-UHFFFAOYSA-N

General description

Atomic number of base material: 50 Tin

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Li et al.
Journal of the American Chemical Society, 136(41), 14589-14597 (2014-09-30)
New electron-acceptor materials are long sought to overcome the small photovoltage, high-cost, poor photochemical stability, and other limitations of fullerene-based organic photovoltaics. However, all known nonfullerene acceptors have so far shown inferior photovoltaic properties compared to fullerene benchmark [6,6]-phenyl-C60-butyric acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service