Skip to Content
Merck
All Photos(1)

Key Documents

366048

Sigma-Aldrich

Benzoyl chloride-d5

99 atom % D

Synonym(s):

Pentadeuterobenzoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$356.00

$356.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$356.00

About This Item

Linear Formula:
C6D5COCl
CAS Number:
Molecular Weight:
145.60
Beilstein:
1955377
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.13

$356.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

protein expression: suitable

refractive index

n20/D 1.552 (lit.)

bp

198 °C (lit.)

mp

-1 °C (lit.)

density

1.254 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(Cl)=O

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

PASDCCFISLVPSO-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Cheng et al.
Combinatorial chemistry & high throughput screening, 20(2), 133-139 (2016-12-22)
Esophageal Squamous Cell Carcinoma (ESCC) is a common malignant tumor in China, which causes about 200,000 deaths each year. Sensitive biomarkers are helpful to diagnose the disease in early stage. To identify biomarkers of ESCC and elucidate underlying mechanism of
Chang Wang et al.
Analytica chimica acta, 1002, 50-61 (2018-01-08)
Quantitative analysis of glycans is an emerging field in glycomic research. Herein we present a rapid and effective dual-labeling strategy, in the combination of isotopic derivatization of N-glycosylamine-based glycans by d0/d5-benzoyl chloride and methylamidation of sialic acids, to relatively quantify
Michael F Wempe et al.
Drug design, development and therapy, 6, 323-339 (2012-11-16)
Human uric acid transporter 1 (hURAT1; SLC22A12) is a very important urate anion exchanger. Elevated urate levels are known to play a pivotal role in cardiovascular diseases, chronic renal disease, diabetes, and hypertension. Therefore, the development of potent uric acid
Fumio Kondo et al.
Analytical and bioanalytical chemistry, 407(18), 5261-5272 (2015-01-31)
Determination of neuroactive substances, such as neurotransmitters and prostanoids, in the extracellular space of the living brain is a very important technique in neuroscience. The hypothalamic paraventricular nucleus (PVN) is one of the most important autonomic control centers in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service