Skip to Content
Merck
All Photos(1)

Key Documents

361976

Sigma-Aldrich

Methyl 1-bromocyclohexanecarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H10CO2CH3
CAS Number:
Molecular Weight:
221.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

115-116 °C/19 mmHg (lit.)

density

1.39 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

COC(=O)C1(Br)CCCCC1

InChI

1S/C8H13BrO2/c1-11-7(10)8(9)5-3-2-4-6-8/h2-6H2,1H3

InChI key

RNWRVPVPLQMTAP-UHFFFAOYSA-N

General description

Methyl 1-bromocyclohexanecarboxylate reacts with zinc and substituted chalcones to yield the corresponding spiro-3,4-dihydropyran-2-one derivatives, 3-aryl-5-aryl-2-oxaspiro[5.5]undec-3-en-1-ones.[1] Reformatsky reaction of methyl 1-bromocyclohexanecarboxylate with 2-aryl-2-oxoacetaldehyde is reported.[2] Methyl 1-bromocyclohexanecarboxylate reacts with zinc and amides or methylamides of 3-aryl-2-cyanopropenoic acids to yield 5-aryl-1,3-dioxo-2-azaspiro[5.5]undecane-4-carbonitriles or 5-aryl-2-methyl-1,3-dioxo-2-azaspiro[5.5]undecane-4-carbonitriles.[3]

Application

Methyl 1-bromocyclohexanecarboxylate may be used in the improved method of preparation of methyl 3-oxo-1-cyclohexene-1-carboxylate.[4] It may be used in the Reformatsky synthesis of 16-aryl-15-oxadispiro[5.1.5.3]hexadecane-7,14-diones.[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirillov NF, et al.
Russ. J. Org. Chem., 40(7), 953-956 (2004)
Improved preparation of methyl 3-oxo-1-cyclohexene-1-carboxylate and its use in the synthesis of substituted 1, 5-cyclodecadienes.
Lange GL and Otulakowski JA.
The Journal of Organic Chemistry, 47(26), 5093-5096 (1982)
Reformatsky Synthesis of 16-Aryl-15-oxadispiro [5.1. 5.3] hexadecane-7, 14-diones.
Kirillov NF and Shchepin VV.
Russ. J. Org. Chem., 37(9), 1223-1224 (2001)
Reaction of Reformatsky reagent prepared from methyl 1-bromocyclohexanecarboxylate and zinc with substituted chalcones.
Kirillov NF and Gavrilov AG.
Russ. J. Org. Chem., 44(7), 963-964 (2008)
Reaction of methyl 1-bromocyclohexanecarboxylate with zinc and 3-aryl-2-cyanopropenoic acids amides.
Kirillov NF, et al.
Russ. J. Gen. Chem., 82(7), 1228-1232 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service