Skip to Content
Merck
All Photos(1)

Key Documents

361828

Sigma-Aldrich

4-Fluorophenethylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
139.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5072 (lit.)

bp

50-52 °C/0.15 mmHg (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

NCCc1ccc(F)cc1

InChI

1S/C8H10FN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5-6,10H2

InChI key

CKLFJWXRWIQYOC-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Application

4-Fluorophenethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives.[1] It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitra Matloobi et al.
Journal of combinatorial chemistry, 9(2), 275-284 (2007-03-14)
An efficient and rapid microwave-assisted solution-phase method for the synthesis of 2-amino-4-arylpyrimidine-5-carboxylic acid derivatives has been developed. The five-step linear protocol involves an initial Biginelli multicomponent reaction leading to dihydropyrimidine-2-thiones which are subsequently S-alkylated with methyl iodide. The resulting 2-methylthiodihydropyrimidines
Ortho-metalated primary amines. 6.1 The first synthesis of six-membered palladacycles from primary amines containing electron-withdrawing substituents: end of the limiting rules of Cope and Friedrich on cyclopalladation of benzyl-and phenethylamines.
Vicente J, et al.
Organometallics, 22(26), 5513-5517 (2003)
Tianqi Niu et al.
Nano letters, 19(10), 7181-7190 (2019-09-04)
Perovskite solar cells based on two-dimensional/three-dimensional (2D/3D) hierarchical structure have attracted significant attention in recent years due to their promising photovoltaic performance and stability. However, obtaining a detailed understanding of interfacial mechanism at the 2D/3D heterojunction, for example, the ligand-chemistry-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service