Skip to Content
Merck
All Photos(1)

Key Documents

359262

Sigma-Aldrich

Poly(4-vinylpyridine hydrochloride)

100-200 mesh, extent of labeling: ~6.5 mmol/g Cl resin loading, 2 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

powder

crosslinking

2 % cross-linked with divinylbenzene

extent of labeling

~6.5 mmol/g Cl resin loading

particle size

100-200 mesh

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service