Skip to Content
Merck
All Photos(1)

Key Documents

359246

Sigma-Aldrich

Trimethyloctadecylammonium bromide

98%

Synonym(s):

C18TAB, TMODAB, Octadecyltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$176.00
50 G
$397.00

$176.00


Available to ship on23 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$176.00
50 G
$397.00

About This Item

Linear Formula:
CH3(CH2)17N(Br)(CH3)3
CAS Number:
Molecular Weight:
392.50
Beilstein:
3574090
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$176.00


Available to ship on23 April 2025Details


Request a Bulk Order

Quality Level

Assay

98%

mp

~250 °C (dec.) (lit.)

SMILES string

C[N+](C)(CCCCCCCCCCCCCCCCCC)C.[Br-]

InChI

1S/C21H46N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(2,3)4;/h5-21H2,1-4H3;1H/q+1;/p-1

InChI key

SZEMGTQCPRNXEG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trimethyloctadecylammonium bromide (C18TAB) is generally used as a surfactant.[1] It can be used in the preparation of polyoxomolybdate-surfactant hybrid layered crystals.[2] It can also be used as a surface modifier in the liquid chromatography.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanocomposites composed of HEUR polymer and magnetite iron oxide nanoparticles: Structure and magnetic response of the hydrogel and dried state.
Campanella A, et al.
Polymer, 60, 176-185 (2015)
Determination of UV-absorbing anions in environmental samples by microcolumn high-performance liquid chromatography.
Vereshchagin A L, et al.
Journal of Analytical Chemistry, 55(10), 1000-1002 (2000)
Layered and molecular-structural control in polyoxomolybdate hybrid crystals by surfactant chain length.
Ito T, et al.
Journal of Molecular Structure, 1106, 220-226 (2016)
Juan C López-Montilla et al.
Water research, 39(9), 1907-1913 (2005-05-19)
The removal of model pollutants bromocresol green (BG) and phenol from water is demonstrated via two liquid-liquid extraction methods. Both methods exploit selective interactions established by the pollutant molecule with a surfactant, oil, or alcohol, and are variants of the
Yutaka Sumino et al.
The journal of physical chemistry. B, 113(48), 15709-15714 (2009-11-12)
Spontaneous deformation of a tetradecane droplet with palmitic acid on an aqueous phase with stearyltrimethylammonium chloride is reported. Palmitic acid is transported from the oil droplet to the aqueous phase by the concentration difference between the organic and the aqueous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service