Skip to Content
Merck
All Photos(1)

Documents

357677

Sigma-Aldrich

2,5-Dimethoxyphenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

refractive index

n20/D 1.5424 (lit.)

bp

160 °C/10 mmHg (lit.)

density

1.089 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC)c(CCN)c1

InChI

1S/C10H15NO2/c1-12-9-3-4-10(13-2)8(7-9)5-6-11/h3-4,7H,5-6,11H2,1-2H3

InChI key

WNCUVUUEJZEATP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dimethoxyphenethylamine is a phenethylamine-type designer drug and is commonly known as psychoactive drug. It was detected as psychostimulant in the urine using liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Application

2,5-Dimethoxyphenethylamine may be used in the preparation of aminoketone hydrochloride.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amines Related to 2, 5-Dimethoxyphenethylamine. 1 II.
Baltzly R and Buck JS.
Journal of the American Chemical Society, 62(1), 164-167 (1940)
Sarah Kerrigan et al.
Journal of forensic sciences, 59(1), 175-183 (2013-12-10)
Designer psychostimulants are known by recreational drug users to produce a complex array of adrenergic and hallucinogenic effects. Many of these drugs are not targeted during routine toxicology testing and as a consequence, they are rarely reported. The purpose of
Simultaneous analysis of six phenethylamine-type designer drugs by TLC, LC-MS, and GC-MS.
Kanai K, et al.
Forensic Toxicology, 26(1), 6-12 (2008)
Eline Pottie et al.
Biochemical pharmacology, 182, 114251-114251 (2020-10-01)
Psychedelic new psychoactive substances (NPS), compounds exerting their main pharmacological effects through the activation of the serotonin 2A receptor (5-HT2AR), continuously comprise a substantial portion of the reported NPS. However, these substances and their exact mechanism of action, differentiating them

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service