Skip to Content
Merck
All Photos(1)

Key Documents

357502

Sigma-Aldrich

2,4-Dichlorophenethylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
190.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.565 (lit.)

bp

75-76 °C/0.04 mmHg (lit.)

density

1.26 g/mL at 25 °C (lit.)

SMILES string

NCCc1ccc(Cl)cc1Cl

InChI

1S/C8H9Cl2N/c9-7-2-1-6(3-4-11)8(10)5-7/h1-2,5H,3-4,11H2

InChI key

VHJKDOLGYMULOP-UHFFFAOYSA-N

Application

2,4-Dichlorophenethylamine may be used in the synthesis of cis-(L-Arginyl)amino-N-(2,4-dichlorophenylmethyl)-L-proline 2,4-dichlorophenethylamide.[1] It may be used in the synthesis of various N-alkylglycine.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Quintanar-Audelo et al.
Journal of medicinal chemistry, 50(24), 6133-6143 (2007-11-08)
The vanilloid receptor subunit 1, or transient receptor potential vanilloid 1 (TRPV1), integrates physical and chemical stimuli in the peripheral nervous system, playing a key role in inflammatory pain. Identification of potent TRPV1 antagonists is thus an important goal of
Carolina García-Martínez et al.
The journal of pain : official journal of the American Pain Society, 7(10), 735-746 (2006-10-05)
Vanilloid receptor subunit 1 (TRPV1) is an integrator of physical and chemical stimuli in the peripheral nervous system. This receptor plays a key role in the pathophysiology of inflammatory pain. Thus, the identification of receptor antagonists with analgesic and anti-inflammatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service