Skip to Content
Merck
All Photos(1)

Key Documents

357383

Sigma-Aldrich

Platinum-iridium alloy 70:30

wire, diam. 0.5 mm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:

form

wire

composition

Iridium, 30 wt. %
Platinum, 70 wt. %

diam.

0.5 mm

SMILES string

[Ir].[Pt]

InChI

1S/Ir.Pt

InChI key

HWLDNSXPUQTBOD-UHFFFAOYSA-N

Quantity

440 mg = 10 cm; 2.2 g = 50 cm

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service