Skip to Content
Merck
All Photos(1)

Key Documents

349615

Sigma-Aldrich

2,2-Diethoxypropionitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(OC2H5)2CN
CAS Number:
Molecular Weight:
143.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

60 °C/20 mmHg (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CCOC(C)(OCC)C#N

InChI

1S/C7H13NO2/c1-4-9-7(3,6-8)10-5-2/h4-5H2,1-3H3

InChI key

QPFCLGADWIWYFW-UHFFFAOYSA-N

General description

2,2-Diethoxypropionitrile can be prepared by the reaction of orthoacetic acid ethyl ester with HCN.

Application

2,2-Diethoxypropionitrile may be used in the synthesis of [2H4]-acetyl-2-thiazoline. It may be used in the synthesis of 2-acetyl-2-thiazoline by condensation reaction with cysteamine, followed by the hydrolysis.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of potent odorants in roasted beef by aroma extract dilution analysis.
Cerny C and Grosch W.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 194(4), 322-325 (1992)
Quantification of character-impact odour compounds of roasted beef.
Cerny C and Grosch W.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 196(5), 417-422 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service