348309
4-Methylphthalic acid
99%
Synonym(s):
4-Methyl-1,2-benzenedicarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H3-1,2-(CO2H)2
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Assay
99%
form
powder
mp
146-148 °C (lit.)
SMILES string
Cc1ccc(C(O)=O)c(c1)C(O)=O
InChI
1S/C9H8O4/c1-5-2-3-6(8(10)11)7(4-5)9(12)13/h2-4H,1H3,(H,10,11)(H,12,13)
InChI key
CWJJAFQCTXFSTA-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C P Saint et al.
FEMS microbiology letters, 57(3), 323-328 (1990-06-01)
Genes involved in 4-methyl-o-phthalate and 4-hydroxy-iso-phthalate catabolism reside on a 226-232 kbp catabolic plasmid termed MOP. This was confirmed by transformation and conjugation into an isogenic heat-cured (MOP-) derivative of the wild-type isolate, identified and termed Pseudomonas cepacia Pc701. Transformation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service