Skip to Content
Merck
All Photos(1)

Key Documents

347329

Sigma-Aldrich

Ethyl (R)-(−)-3-hydroxybutyrate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$39.50
5 G
$127.00

$39.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$39.50
5 G
$127.00

About This Item

Linear Formula:
CH3CH(OH)CH2CO2C2H5
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$39.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

optical activity

[α]20/D −46°, c = 1 in chloroform

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.42 (lit.)

bp

75-76 °C/12 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CCOC(=O)C[C@@H](C)O

InChI

1S/C6H12O3/c1-3-9-6(8)4-5(2)7/h5,7H,3-4H2,1-2H3/t5-/m1/s1

InChI key

OMSUIQOIVADKIM-RXMQYKEDSA-N

General description

Ethyl (R)-(-)-3-hydroxybutyrate is a chiral building block for the preparation of bioactive compounds used in the pharmaceutical industry. It is formed during the hydrolysis of poly-3-hydroxybutyrate.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A two-step enzymatic resolution process for large-scale production of (S)-and (R)-ethyl-3-hydroxybutyrate.
Fishman A, et al.
Biotechnology and Bioengineering, 74(3), 256-263 (2001)
Young-Min Han et al.
Molecular cell, 71(6), 1064-1078 (2018-09-11)
β-hydroxybutyrate (β-HB) elevation during fasting or caloric restriction is believed to induce anti-aging effects and alleviate aging-related neurodegeneration. However, whether β-HB alters the senescence pathway in vascular cells remains unknown. Here we report that β-HB promotes vascular cell quiescence, which
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service