Skip to Content
Merck
All Photos(1)

Key Documents

344699

Sigma-Aldrich

Copper(II) pyrophosphate hydrate

Synonym(s):

Cupric pyrophosphate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$133.00

$133.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 G
$133.00

About This Item

Empirical Formula (Hill Notation):
Cu2P2O7 · xH2O
CAS Number:
Molecular Weight:
301.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks
solubility:
H2O: insoluble(lit.)

$133.00


In StockDetails


Request a Bulk Order

form

powder and chunks

Quality Level

composition

Degree of hydration, ~3

concentration

33-35% Cu (EDTA titration)

solubility

H2O: insoluble(lit.)

application(s)

battery manufacturing

SMILES string

O.[Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/2Cu.H4O7P2.H2O/c;;1-8(2,3)7-9(4,5)6;/h;;(H2,1,2,3)(H2,4,5,6);1H2/q2*+2;;/p-4

InChI key

PXZSJEBZWADSIY-UHFFFAOYSA-J

Application

Copper(II) pyrophosphate hydrate was one of the constituents in the preparation of an electrolyte which was used to study the application of nickel-tin (Ni-Sn) nanowires as electrodes for Li-ion batteries. [1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stable high areal capacity lithium-ion battery anodes based on three-dimensional. Ni-Sn nanowire networks.
Tian M, et al.
Journal of Power Sources, 211, 46-51 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service