Skip to Content
Merck
All Photos(1)

Key Documents

344257

Sigma-Aldrich

Choline hydroxide solution

20 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2N(CH3)3OH
CAS Number:
Molecular Weight:
121.18
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

liquid

Quality Level

concentration

20 wt. % in H2O

refractive index

n20/D 1.37

density

1.023 g/mL at 25 °C

SMILES string

[OH-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.H2O/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H2/q+1;/p-1

InChI key

KIZQNNOULOCVDM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Dan Hou et al.
Biotechnology and bioengineering, 112(1), 65-73 (2014-07-30)
In this work, 28 cholinium ionic liquids (ILs), most of which are good solvents for dissolving lignin, were used for rice straw pretreatment to improve subsequent enzymatic hydrolysis. The anion exerted a significant effect on the pretreatment effectiveness of the
Kyung-Eui Park et al.
Bioscience, biotechnology, and biochemistry, 78(6), 981-988 (2014-07-19)
Phospholipase D (PLD) catalyzes the hydrolysis of phosphatidylcholine (PC), the most abundant phospholipids of plasma membrane, resulting in the production of choline and phosphatidic acid (PA). Choline is a precursor of the neurotransmitter acetylcholine, whereas PA functions as an intracellular
Yan-Xia An et al.
Bioresource technology, 192, 165-171 (2015-06-01)
Pretreatment of lignocelluloses is a key step in the biorefinery for production of biofuels and valuable platform chemicals. In this work, various lignocelluloses were pretreated using cholinium ionic liquids (ILs) that are wholly composed of biomaterials, and fractionated into carbohydrate-rich
Bieke Onghena et al.
Dalton transactions (Cambridge, England : 2003), 43(30), 11566-11578 (2014-06-19)
The ionic liquid choline bis(trifluoromethylsulfonyl)imide, [Chol][Tf2N], was used for the extraction of neodymium(III), in combination with choline hexafluoroacetylacetonate, [Chol][hfac], as the extractant. The binary mixture of [Chol][Tf2N] and water shows temperature-dependent phase behavior, with an upper critical solution temperature of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service