Skip to Content
Merck
All Photos(1)

Key Documents

343781

Sigma-Aldrich

Maleic anhydride-d2

98 atom % D

Synonym(s):

2,5-Furandione-3,4-d2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$463.00
5 G
$1,330.00

$463.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$463.00
5 G
$1,330.00

About This Item

Empirical Formula (Hill Notation):
C4D2O3
CAS Number:
Molecular Weight:
100.07
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$463.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

form

powder

bp

200 °C (lit.)

mp

51-56 °C (lit.)

mass shift

M+2

SMILES string

[2H]C1=C([2H])C(=O)OC1=O

InChI

1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H/i1D,2D

InChI key

FPYJFEHAWHCUMM-QDNHWIQGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service