Skip to Content
Merck
All Photos(2)

Key Documents

343587

Sigma-Aldrich

Methyl 4,4-dimethoxy-3-oxovalerate

97%

Synonym(s):

Methyl 4,4-dimethoxy-3-oxopentanoate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$492.00

$492.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$492.00

About This Item

Linear Formula:
CH3C(OCH3)2COCH2CO2CH3
CAS Number:
Molecular Weight:
190.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$492.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

69-73 °C/0.35 mmHg (lit.)

density

1.111 g/mL at 25 °C (lit.)

functional group

ester
ether
ketal
ketone

SMILES string

COC(=O)CC(=O)C(C)(OC)OC

InChI

1S/C8H14O5/c1-8(12-3,13-4)6(9)5-7(10)11-2/h5H2,1-4H3

InChI key

ZQFJOBUKEHFXKY-UHFFFAOYSA-N

General description

Methyl 4,4-dimethoxy-3-oxovalerate is an ester. Preparation of methyl 4,4-dimethoxy-3-oxovalerate in tetrahydrofuran is reported.[1]

Application

Methyl 4,4-dimethoxy-3-oxovalerate may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereochemical control in the synthesis of the cyclohexyl portion of the milbemycin skeleton.
Turnbull MD, et al.
Tetrahedron Letters, 25(47), 5449-5452 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service