Skip to Content
Merck
All Photos(2)

Documents

343390

Sigma-Aldrich

1-Cyano-1-cyclopropanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC3H4CO2H
CAS Number:
Molecular Weight:
111.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

140 °C (lit.)

SMILES string

OC(=O)C1(CC1)C#N

InChI

1S/C5H5NO2/c6-3-5(1-2-5)4(7)8/h1-2H2,(H,7,8)

InChI key

KSJJMSKNZVXAND-UHFFFAOYSA-N

General description

1-Cyano-1-cyclopropanecarboxylic acid (1-Cyanocyclopropanecarboxylic acid) is a substituted cyclopropane. It is the intermediate formed during homoconjugate addition of nucleophiles to cyclopropane-1,1-dicarboxylate derivative.

Application

1-Cyano-1-cyclopropanecarboxylic acid may be used in the synthesis of amidine-based nanomolar sphingosine kinase1 subtype-selective inhibitors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homoconjugate Addition of Nucleophiles to Cyclopropane-1, 1-Dicarboxylate Derivatives: 2-Oxo-1-Phenyl-3-Pyrrolidinecarboxylic Acid.
Singh RK and Danishefsky S.
Organic Syntheses, 66-66 null
Substituted cyclopropanes. 5. 1-Cyanocyclopropanecarboxylic acid.
Jones PG and Schrumpf G.
Acta Crystallographica Section C, Structural Chemistry, 43(8), 1576-1579 (1987)
Andrew J Kennedy et al.
Journal of medicinal chemistry, 54(10), 3524-3548 (2011-04-19)
Sphingosine 1-phosphate (S1P) is a bioactive lipid that has been identified as an accelerant of cancer progression. The sphingosine kinases (SphKs) are the sole producers of S1P, and thus, SphK inhibitors may prove effective in cancer mitigation and chemosensitization. Of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service