Skip to Content
Merck
All Photos(3)

Documents

342203

Sigma-Aldrich

Strontium fluoride

powder, <5 μm

Synonym(s):

Strontium difluoride (SrF2 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrF2
CAS Number:
Molecular Weight:
125.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

concentration

66.2-73.2% Sr (EDTA titration)

particle size

<5 μm

bp

2489 °C (lit.)

mp

>1400 °C (lit.)

density

4.24 g/mL at 25 °C (lit.)

SMILES string

F[Sr]F

InChI

1S/2FH.Sr/h2*1H;/q;;+2/p-2

InChI key

FVRNDBHWWSPNOM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bikramjit Basu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(6), 1168-1179 (2014-10-11)
One of the desired properties for any new biomaterial composition is its long-term stability in a suitable animal model and such property cannot be appropriately assessed by performing short-term implantation studies. While hydroxyapatite (HA) or bioglass coated metallic biomaterials are
Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service