Skip to Content
Merck
All Photos(1)

Key Documents

340677

Sigma-Aldrich

Lithium phenylacetylide solution

1.0 M in THF

Synonym(s):

(Phenylethynyl)lithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CLi
CAS Number:
Molecular Weight:
108.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF

density

0.91 g/mL at 25 °C

functional group

phenyl

SMILES string

[Li]C#Cc1ccccc1

InChI

1S/C8H5.Li/c1-2-8-6-4-3-5-7-8;/h3-7H;

InChI key

QQONVLVTCMNCTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service