Skip to Content
Merck
All Photos(1)

Key Documents

340472

Sigma-Aldrich

2,3,4-Trifluorotoluene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$213.00

$213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$213.00

About This Item

Linear Formula:
CH3C6H2F3
CAS Number:
Molecular Weight:
146.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$213.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

126-127 °C (lit.)

density

1.222 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1ccc(F)c(F)c1F

InChI

1S/C7H5F3/c1-4-2-3-5(8)7(10)6(4)9/h2-3H,1H3

InChI key

LRQPEHJWTXCLQY-UHFFFAOYSA-N

General description

Corona discharge of 2,3,4-trifluorotoluene generates difluorobenzyl radicals, which were investigated by vibronically well-resolved emission spectrum.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of Difluorobenzyl Radicals from 2, 3, 4-Trifluorotoluene in Corona Excitation.
Yoon YW, et al.
Bull. Korean Chem. Soc., 32(6), 1993-1996 (2011)
Yangming Lin et al.
ChemSusChem, 10(17), 3497-3505 (2017-07-01)
Selective oxidation of alcohols to aldehydes is widely applicable to the synthesis of various green chemicals. The poor chemoselectivity for complicated primary aldehydes over state-of-the-art metal-free or metal-based catalysts represents a major obstacle for industrial application. Bucky nanodiamond is a
Thi Hai Quyen Nguyen et al.
Physical chemistry chemical physics : PCCP, 22(15), 7699-7709 (2020-03-07)
The transport of both electrons and ions in organic mixed ionic and electronic conductors such as phthalocyanines, is essential to allow redox reactions of entire films and, hence, to impart electrochromism. Thin films of a new type, tetrakis-perfluoroisopropyl-perfluoro phthalocyanine, F40PcCu
Hee-Jin Yoo et al.
Analytical methods : advancing methods and applications, 12(48), 5767-5800 (2020-11-27)
A great deal of effort has been expended to develop accurate means of determining the properties of synthetic polymers using matrix-assisted laser desorption/ionization (MALDI) time-of-flight (TOF) mass spectrometry (MS). Many studies have focused on the importance of sample pre-treatment to
Simon Hammann et al.
Analytical and bioanalytical chemistry, 407(30), 9019-9028 (2015-10-07)
Triacylglycerols represent the major part (>90%) in most plant oils and have to be eliminated, when the minor compounds such as phytosterols or tocopherols should be analyzed. Here, we used an all liquid-liquid chromatographic technique, countercurrent chromatography (CCC), to fractionate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service