Skip to Content
Merck
All Photos(2)

Key Documents

339644

Sigma-Aldrich

Triethoxymethylsilane

technical grade, 90%

Synonym(s):

Methyltriethoxysilane

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$89.40

$89.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$89.40

About This Item

Linear Formula:
CH3Si(OC2H5)3
CAS Number:
Molecular Weight:
178.30
Beilstein:
1742453
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$89.40


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

vapor density

>1 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

90%

form

liquid

refractive index

n20/D 1.383 (lit.)

bp

141-143 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(OCC)OCC

InChI

1S/C7H18O3Si/c1-5-8-11(4,9-6-2)10-7-3/h5-7H2,1-4H3

InChI key

CPUDPFPXCZDNGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that

Articles

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service