Skip to Content
Merck
All Photos(2)

Key Documents

339555

Sigma-Aldrich

Bromomethyl phenyl sulfone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2Br
CAS Number:
Molecular Weight:
235.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

51-53 °C (lit.)

functional group

bromo
sulfone

SMILES string

BrCS(=O)(=O)c1ccccc1

InChI

1S/C7H7BrO2S/c8-6-11(9,10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

SKIMEKUYIQHJQV-UHFFFAOYSA-N

General description

Reaction of bromomethyl phenyl sulfone with potassium thiophenoxide in dimethyl formamide gave phenylthiomethyl phenyl sulfone.[1] It undergoes disproportionation with KOH in DMSO at room temperature to yield methyl phenyl sulfone and dibromomethyl phenyl sulfone.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleophilic displacements alpha to sulfonyl groupings.
Bordwell FG and Jarvis BB.
The Journal of Organic Chemistry, 33(3), 1182-1185 (1968)
Base-induced disproportionation of halomethyl phenyl sulfones to methyl and dihalomethyl phenyl sulfones.
Galvagni M, et al.
The Journal of Organic Chemistry, 55(14), 4454-4456 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service