Skip to Content
Merck
All Photos(1)

Documents

338966

Sigma-Aldrich

3,8-Diamino-6-phenylphenanthridine

98%

Synonym(s):

6-Phenyl-3,8-phenanthridinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N3
CAS Number:
Molecular Weight:
285.34
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

impurities

~2% ammonium chloride

mp

196-198 °C (lit.)

SMILES string

Nc1ccc2c(c1)nc(-c3ccccc3)c4cc(N)ccc24

InChI

1S/C19H15N3/c20-13-6-8-15-16-9-7-14(21)11-18(16)22-19(17(15)10-13)12-4-2-1-3-5-12/h1-11H,20-21H2

InChI key

CPNAVTYCORRLMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of rigid polyamides. It has been investigated for its anti-tumor and anti-viral properties. Small organic molecules, such as 3,8-Diamino-6-phenylphenanthridine, can bind to DNA by means of a non-specific binding along the DNA exterior.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatic polyamides of 3, 8-diamino-6-phenylphenanthridine and their molecular complexes with sulfonated polystyrene ionomers.
Cimecioglu AL, et al.
Macromolecules, 28(18), 6343-6346 (1995)
On the electronic structure of ethidium.
Luedtke NW, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 11(2), 495-508 (2005)
Binding of cationic and neutral phenanthridine Intercalators to a DNA oligomer is controlled by dispersion energy: Quantum chemical calculations and molecular mechanics simulations.
Kubar T, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 12(1), 280-290 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service