Skip to Content
Merck
All Photos(1)

Key Documents

338958

Sigma-Aldrich

Triethyl 3-methyl-4-phosphono-2-butenoate, mixture of cis and trans

technical grade, 80%

Synonym(s):

Diethyl 3-ethoxycarbonyl-2-methyl-2-propenyl phosphonate, Triethyl 3-methyl-4-phosphonocrotonate, Triethyl 4-phosphonosenecioate, mixture of cis and trans

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$147.00

$147.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$147.00

About This Item

Linear Formula:
(C2H5O)2P(O)CH2C(CH3)=CHCO2C2H5
CAS Number:
Molecular Weight:
264.26
Beilstein:
2417880
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$147.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

technical grade

Assay

80%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.46 (lit.)

density

1.089 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\C=C(/C)CP(=O)(OCC)OCC

InChI

1S/C11H21O5P/c1-5-14-11(12)8-10(4)9-17(13,15-6-2)16-7-3/h8H,5-7,9H2,1-4H3/b10-8+

InChI key

OQKGPUSERILONW-CSKARUKUSA-N

Application

Reactant for preparation of:
  • Bicyclodecenes and oxabicyclodecenes via intramolecular [4 + 3]-cycloaddition, using alkylation, olefination and coupling
  • Methyloctatrienoic acid retinoid X receptor modulators from insulin sensitizers for the treatment of type II diabetes

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service