Skip to Content
Merck
All Photos(1)

Key Documents

338648

Sigma-Aldrich

4′-Hexyl-4-biphenylcarbonitrile

liquid crystal (nematic), 98%

Synonym(s):

4′-n -Hexyl-4-cyanobiphenyl, 4′-Hexyl-4-biphenylcarbonitrile, 4′-Hexyl-4-cyanobiphenyl, 4′-Hexyl[1,1′-biphenyl]-4-carbonitrile, 4-Cyano-4′-n -hexylbiphenyl, 4-Cyano-4′-hexyl-1,1′-biphenyl, 4-Cyano-4′-hexylbiphenyl, 4-Hexyl-4′-cyanobiphenyl, 4-Hexyl-4′-cyanodiphenyl, p -Hexyl-p ′-cyanobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H4C6H4CN
CAS Number:
Molecular Weight:
263.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid crystal (nematic)

SMILES string

CCCCCCc1ccc(cc1)-c2ccc(cc2)C#N

InChI

1S/C19H21N/c1-2-3-4-5-6-16-7-11-18(12-8-16)19-13-9-17(15-20)10-14-19/h7-14H,2-6H2,1H3

InChI key

VADSDVGLFDVIMG-UHFFFAOYSA-N

General description

4′-Hexyl-4-biphenylcarbonitrile belongs to the class of low molecular weight liquid crystals (LCs). Optical properties of LCs can be controlled thermally. It can be rendered opaque to transparent state by thermal switching. This effect appears on account of birefringence feature of LCs. Low molecular weight LCs show high, and strongly dependence on temperature as well as possesses higher solubility in polymer matrices. It also exhibits a strong dipole moment.

Application

  • Used in thermoresponsive meso/nanostructured PS-b-PEO block copolymer-dispersed liquid crystal.
  • 4′-hexyl-4-biphenylcarbonitrile, 4′-octyloxy-4-biphenylcarbonitrile and their composite liquid crystals were studied for dielectric properties.
Proposed applications are in the field of
  • thermo- and electro optical devices (e.g. optical shutters, smart windows, optical sensors,memories and flexible display devices),
  • n phase array optics (e.g. as switchable birefringent phase shifters).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermoresponsive meso/nanostructured thermosetting materials based on PS-b-PEO block copolymer-dispersed liquid crystal: Curing behavior and morphological variation.
Tercjak A, et al.
Acta Materialia, 56(18), 5112-5122 (2008)
Conductance and Dielectric Anisotropy Properties of 4'-Hexyl-4-biphenylcarbonitrile and 4'-Octyloxy-4-biphenylcarbonitrile Liquid Crystals and Their Composite.
Ozgan S, et al.
Asian Journal of Research in Chemistry, 23(7), 3247-3251 (2011)
Hiroshi Kakiuchida et al.
ACS applied materials & interfaces, 11(21), 19404-19412 (2019-05-08)
A simple nonuniform irradiation method for photopolymerization-induced phase separation (PPIPS) was developed to produce unconventional mesoscale domain structures composed of liquid crystal (LC) and reactive mesogen (RM) phases. The LC/RM phase formations and their molecular orientation ordering through PPIPS were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service