Skip to Content
Merck
All Photos(1)

Key Documents

338109

Sigma-Aldrich

N-(Phosphonomethyl)glycine, monoisopropylamine salt solution

40 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)CH2NHCH2CO2H·(CH3)2CHNH2
CAS Number:
Molecular Weight:
228.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

40 wt. % in H2O

refractive index

n20/D 1.435

density

1.218 g/mL at 25 °C

SMILES string

CC(C)N.OC(=O)CNCP(O)(O)=O

InChI

1S/C3H8NO5P.C3H9N/c5-3(6)1-4-2-10(7,8)9;1-3(2)4/h4H,1-2H2,(H,5,6)(H2,7,8,9);3H,4H2,1-2H3

InChI key

ZEKANFGSDXODPD-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fate and Mobility of Glyphosate Leachate in Palestinian Soil Using Soil Column.
Jodeh S, et al.
Journal of Materials and Environmental Science, 5(6), 2008-2016 (2014)
In situ fluorimetric determination of pesticides on vegetables.
Hassoon S and Schechter I.
Analytica Chimica Acta, 405(1), 9-15 (2000)
Lee H-L and Guo H-R.
Herbicides, Theory and Applications, 978-953 (2011)
Dimitra Papagiannaki et al.
Scientific reports, 10(1), 20247-20247 (2020-11-22)
The active herbicide ingredient glyphosate [N-(phosphonomethyl)glycine] is frequently detected as a contaminant in groundwater and surface waters. This study investigated effects of UV-A (365 nm), UV-B (302 nm) and UV-C (254 nm) irradiation of glyphosate in water on photolysis and toxicity to aquatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service