337749
Benzo[a]anthracene-7,12-dione
purified by sublimation, 98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H10O2
CAS Number:
Molecular Weight:
258.27
Beilstein:
2052941
EC Number:
MDL number:
UNSPSC Code:
12352100
Assay
98%
purified by
sublimation
mp
169-171 °C (lit.)
SMILES string
O=C1c2ccccc2C(=O)c3c1ccc4ccccc34
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service