Skip to Content
Merck
All Photos(1)

Key Documents

337595

Sigma-Aldrich

(S)-(−)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone

99%

Synonym(s):

(S)-(−)-1-(tert-Butoxycarbonyl)-2-tert-butyl-3-methyl-4-imidazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$261.00
1 G
$1,000.00

$261.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
250 MG
$261.00
1 G
$1,000.00

About This Item

Empirical Formula (Hill Notation):
C13H24N2O3
CAS Number:
Molecular Weight:
256.34
Beilstein:
4907144
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$261.00


0 left in stock

Request a Bulk Order

Assay

99%

optical activity

[α]25/D −14°, c = 1 in methylene chloride

optical purity

ee: 99% (GLC)

mp

67-69 °C (lit.)

application(s)

peptide synthesis

SMILES string

CN1[C@@H](N(CC1=O)C(=O)OC(C)(C)C)C(C)(C)C

InChI

1S/C13H24N2O3/c1-12(2,3)10-14(7)9(16)8-15(10)11(17)18-13(4,5)6/h10H,8H2,1-7H3/t10-/m0/s1

InChI key

HJJLVATZPPJBNG-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service