Skip to Content
Merck
All Photos(3)

Key Documents

336262

Sigma-Aldrich

Sodium stannate trihydrate

95%

Synonym(s):

Sodium stannate hydrate (Na2 SnO3 .3H2O), Sodium tin oxide (Na2 SnO3 ), trihydrate, Sodium tin oxide trihydrate (Na2 SnO3 .3H2O)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$62.80
500 G
$226.00

$62.80


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$62.80
500 G
$226.00

About This Item

Linear Formula:
Na2SnO3·3H2O
CAS Number:
Molecular Weight:
266.73
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
95%
form:
powder

$62.80


In StockDetails


Request a Bulk Order

Quality Level

Assay

95%

form

powder

reaction suitability

reagent type: catalyst
core: tin

SMILES string

O.O.O.[Na+].[Na+].[O-][Sn]([O-])=O

InChI

1S/2Na.3H2O.3O.Sn/h;;3*1H2;;;;/q2*+1;;;;;2*-1;

InChI key

SFXJSNATBHJIDS-UHFFFAOYSA-N

Application

Sodium stannate trihydrate can be used as a precursor to synthesize:
  • Lanthanum stannate nanoparticles that are used for the modification of glassy carbon electrodes.
  • Neodymium stannate nanoparticles for the selective detection of dimetridazole in biological samples.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service