Skip to Content
Merck
All Photos(1)

Documents

333581

Sigma-Aldrich

1,4-Naphthalenedicarboxylic acid

94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CO2H)2
CAS Number:
Molecular Weight:
216.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

94%

form

solid

mp

>300 °C (lit.)

SMILES string

OC(=O)c1ccc(C(O)=O)c2ccccc12

InChI

1S/C12H8O4/c13-11(14)9-5-6-10(12(15)16)8-4-2-1-3-7(8)9/h1-6H,(H,13,14)(H,15,16)

InChI key

ABMFBCRYHDZLRD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Liu et al.
Chemistry, an Asian journal, 14(12), 2072-2076 (2019-05-09)
In this study, we report a facile ligand-assisted in situ hydrothermal approach for preparation of compact [Al(OH)(1,4-NDC)] (1,4-NDC=1,4-naphthalenedicarboxylate) MOF membranes on porous γ-Al2 O3 substrates, which also served as the Al3+ source of MOF membranes. Simultaneously, it was observed that
Paul D Greenstein et al.
The journal of physical chemistry. B, 121(19), 5086-5093 (2017-05-11)
Using proton and carbon chemical shifts, we investigated the self-association of three isomers of naphthalenedicarboxylic acid, a model for the aggregation of asphaltenes. Experimental proton chemical shifts of each isomer were measured as a function of concentration in an aprotic
Gargi Dutta et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(37), 8932-8940 (2017-04-20)
Three new metal-organic porphyrinic framework compounds of zinc and 5,10,15,20-tetrakis(4-pyridyl)porphyrin (TPyP) have been synthesized under solvothermal conditions. The compounds [Zn
P Arul et al.
Mikrochimica acta, 187(12), 650-650 (2020-11-10)
A copper-1,4-naphthalenedicarboxylic acid-based organic framework (Cu-NDCA MOF) with different morphologies was synthesized by solvothermal synthetic route via a simple protonation-deprotonation approach. The synthesized Cu-NDCA MOFs were analyzed by diverse microscopic and spectral techniques. The FE-SEM and TEM image results exhibited
Timo Rabe et al.
Dalton transactions (Cambridge, England : 2003), 49(15), 4861-4868 (2020-03-29)
Metal-organic frameworks containing Ga3+ ions and four differently substituted naphthalenedicarboxylates (ndc2-) have been synthesized and characterized. The Ga3+ ions are six-fold coordinated by oxygen atoms in all title compounds, but two different inorganic building units, i.e. trans corner-sharing and cis,trans

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service