Skip to Content
Merck
All Photos(2)

Key Documents

333328

Sigma-Aldrich

6-Ethoxy-2-benzothiazolesulfonamide

97%

Synonym(s):

6-Ethoxyzolamide, Ethoxzolamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2O3S2
CAS Number:
Molecular Weight:
258.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

190-193 °C (lit.)

SMILES string

CCOc1ccc2nc(sc2c1)S(N)(=O)=O

InChI

1S/C9H10N2O3S2/c1-2-14-6-3-4-7-8(5-6)15-9(11-7)16(10,12)13/h3-5H,2H2,1H3,(H2,10,12,13)

InChI key

OUZWUKMCLIBBOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Ethoxy-2-benzothiazolesulfonamide is a membrane permeable carbonic anhydrase inhibitor and its ocular disposition and pharmacology in normal albino rabbits has been investigated.[1] It causes inhibition of insulin degradation in vitro.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaspar Peña-Münzenmayer et al.
Pflugers Archiv : European journal of physiology, 466(7), 1317-1327 (2013-10-02)
TASK-2 is a K2P K(+) channel considered as a candidate to mediate CO2 sensing in central chemosensory neurons in mouse. Neuroepithelial cells in zebrafish gills sense CO2 levels through an unidentified K2P K(+) channel. We have now obtained zfTASK-2 from
Andreza Ribeiro et al.
Journal of the Royal Society, Interface, 9(74), 2059-2069 (2012-04-12)
Polymeric micelles of single and mixed poloxamines (Tetronic) were evaluated regarding their ability to host the antiglaucoma agent ethoxzolamide (ETOX) for topical ocular application. Three highly hydrophilic varieties of poloxamine (T908, T1107 and T1307) and a medium hydrophilic variety (T904)
Larissa A Shimoda et al.
American journal of physiology. Lung cellular and molecular physiology, 292(4), L1002-L1012 (2007-01-09)
Hypoxic pulmonary vasoconstriction (HPV) occurs with ascent to high altitude and can contribute to development of high altitude pulmonary edema (HAPE). Vascular smooth muscle contains carbonic anhydrase (CA), and acetazolamide (AZ), a CA inhibitor, blunts HPV and might be useful
Matthias C Jecklin et al.
Journal of molecular recognition : JMR, 22(4), 319-329 (2009-04-18)
We performed a systematic comparison of three label-free methods for quantitative assessment of binding strengths of proteins interacting with small molecule ligands. The performance of (1) nanoelectrospray ionization mass spectrometry (nESI-MS), (2) surface plasmon resonance (SPR), and (3) isothermal titration
T H Maren et al.
Experimental eye research, 55(1), 73-79 (1992-07-01)
We have measured sequentially the concentrations of ethoxzolamide (6-ethoxybenzothiazole-2-sulfonamide) in ocular tissues following its intravenous or topical administration to normal albino rabbits. This was done in parallel with determinations of intraocular pressure (IOP) measured by tonometer or direct manometry. Ethoxzolamide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service