Skip to Content
Merck
All Photos(1)

Key Documents

333301

Sigma-Aldrich

2-Methyl-3-indoleacetic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$161.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$161.00

About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

205 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

Cc1[nH]c2ccccc2c1CC(O)=O

InChI

1S/C11H11NO2/c1-7-9(6-11(13)14)8-4-2-3-5-10(8)12-7/h2-5,12H,6H2,1H3,(H,13,14)

InChI key

QJNNHJVSQUUHHE-UHFFFAOYSA-N

General description

2-Methyl-3-indoleacetic acid is a common plant growth hormone.[1] Relative activity of 2-methyl-3-indoleacetic acid in promoting growth (elongation) in plant cells was evaluated.[2]

Application

  • Reactant for preparation of N-methyl-N-[[(nitrophenyl)sulfonyl]oxy]indoleacetamide derivatives
  • Reactant for preparation of indomethacin analogs and derivatives
  • Reactant for coupling reaction with diazomethane
  • Reactant for prepnaration of (methylindolyl)methyltriazolethiones and (methylindolyl)methylthiadiazolylamines as antidepressant/anticonvulsant agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the Mechanism of Action of Growth Regulators.
R M Muir et al.
Plant physiology, 28(2), 218-232 (1953-04-01)
Advances in the catalytic production of valuable levulinic acid derivatives.
Zhang J, et al.
ChemCatChem, 4(9), 1230-1237 (2012)
Zi-Ao Huang et al.
Electrophoresis, 41(3-4), 183-193 (2019-12-19)
In this paper, the development of a simple dilute-and-shoot method for quantifying urinary creatinine by CE-ESI-MS was described. The creatinine analysis time was about 7 min/sample by conventional single injection (SI) method and can be significantly reduced to less than 2 min/sample

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service