Skip to Content
Merck
All Photos(2)

Key Documents

332186

Sigma-Aldrich

1-Octyl-2-pyrrolidone

98%

Synonym(s):

1-Octyl-2-pyrrolidinone, N-Octyl-2-pyrrolidinone, N-Octyl-2-pyrrolidone, N-Octylbutyrolactam, N-Octylpyrrolidinone, N-Octylpyrrolidone, N-n-Octyl-α-pyrrolidone, N-n-Octyl-2-pyrrolidone

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$131.00
1 L
$448.00

$131.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
$131.00
1 L
$448.00

About This Item

Empirical Formula (Hill Notation):
C12H23NO
CAS Number:
Molecular Weight:
197.32
Beilstein:
1526318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$131.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.465 (lit.)

bp

170-172 °C/15 mmHg (lit.)

mp

−25 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN1CCCC1=O

InChI

1S/C12H23NO/c1-2-3-4-5-6-7-10-13-11-8-9-12(13)14/h2-11H2,1H3

InChI key

WPPOGHDFAVQKLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octyl-2-pyrrolidone is a permeation enhancer and its effect in transport of steroidal permeants across hairless mouse skin was investigated via a parallel pathway skin model.[1]

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yoneto et al.
Journal of pharmaceutical sciences, 87(2), 209-214 (1998-03-31)
In a previous study, the enhancing effects of a series of 1-alkyl-2-pyrrolidones (APs; 1-ethyl, 1-butyl, 1-hexyl, and 1-octyl-2-pyrrolidone) on the transport of steroidal permeants across hairless mouse skin were investigated via a parallel pathway skin model. Isoenhancement concentration conditions were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service